Assay Details
Target Gene Details
Entrez Gene ID: | 115286 |
Gene Name: | solute carrier family 25 member 26 |
Gene Aliases: |
COXPD28, SAMC |
Location: |
Chr.3:66133610-66380021 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC25A26 | NM_173471.3 | NP_775742.4 | ||
AK092495.1 | ||||
AX747571.1 | ||||
DA845619.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv590546 | Chr.3:65487766 - 67232113 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv3596435 | Chr.3:66149175 - 66174448 on Build GRCh38 | Gain |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)