Assay Details
Target Gene Details
Entrez Gene ID: | 7047 |
Gene Name: | transglutaminase 4 |
Gene Aliases: |
TGP, hTGP |
Location: |
Chr.3:44874592-44914601 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TGM4 | NM_003241.3 | NP_003232.2 | ||
XM_011534042.1 | XP_011532344.1 | |||
AK125536.1 | ||||
AK300523.1 | ||||
AK312936.1 | ||||
BC007003.1 | AAH07003.1 | |||
DA870110.1 | ||||
L34840.1 | AAB95430.1 | |||
U31905.1 | AAC50516.1 | |||
U79008.1 | AAB38324.1 | |||
Y09559.1 | CAA70753.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834675 | Chr.3:44805220 - 44966381 on Build GRCh38 | Gain | TGM4 TMEM42 MIR564 KIF15 ZDHHC3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |