Assay Details
Target Gene Details
Entrez Gene ID: | 10533 |
Gene Name: | autophagy related 7 |
Gene Aliases: |
APG7-LIKE, APG7L, GSA7 |
Location: |
Chr.3:11272324-11564704 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv436362 | Chr.3:10048344 - 11860564 on Build GRCh38 | Deletion | FANCD2OS SLC6A1 GHRL LINC00606 ATP2B2-IT2 VHL LINC00852 GHRLOS FANCD2 MIR378B IRAK2 BRK1 SEC13 VGLL4 SLC6A11 SLC6A1-AS1 MIR885 ATP2B2 ATG7 HRH1 LOC101927467 TAMM41 TATDN2 |
dgv762e201 | Chr.3:10008967 - 11913136 on Build GRCh38 | Deletion | GHRL LINC00606 ATP2B2-IT2 GHRLOS CIDECP SEC13 SLC6A1-AS1 ATP2B2 LOC101927467 TAMM41 FANCD2OS EMC3 SLC6A1 LOC401052 VHL LINC00852 FANCD2 MIR378B IRAK2 BRK1 VGLL4 SLC6A11 MIR885 FANCD2P2 ATG7 HRH1 TATDN2 |
nsv1121603 | Chr.3:10071433 - 11889279 on Build GRCh38 | Deletion | FANCD2OS SLC6A1 GHRL LINC00606 ATP2B2-IT2 VHL LINC00852 GHRLOS FANCD2 MIR378B IRAK2 BRK1 SEC13 VGLL4 SLC6A11 SLC6A1-AS1 MIR885 ATP2B2 FANCD2P2 ATG7 HRH1 LOC101927467 TAMM41 TATDN2 |
nsv436869 | Chr.3:10064456 - 11877985 on Build GRCh38 | Insertion | FANCD2OS SLC6A1 GHRL LINC00606 ATP2B2-IT2 VHL LINC00852 GHRLOS FANCD2 MIR378B IRAK2 BRK1 SEC13 VGLL4 SLC6A11 SLC6A1-AS1 MIR885 ATP2B2 FANCD2P2 ATG7 HRH1 LOC101927467 TAMM41 TATDN2 |
nsv1073204 | Chr.3:9996548 - 11875005 on Build GRCh38 | Deletion | EMC3-AS1 GHRL LINC00606 ATP2B2-IT2 GHRLOS CIDECP SEC13 SLC6A1-AS1 ATP2B2 LOC101927467 TAMM41 FANCD2OS EMC3 SLC6A1 LOC401052 VHL LINC00852 FANCD2 MIR378B IRAK2 BRK1 VGLL4 SLC6A11 MIR885 FANCD2P2 ATG7 HRH1 TATDN2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |