Assay Details
Target Gene Details
Entrez Gene ID: | 22907 |
Gene Name: | DEAH-box helicase 30 |
Gene Aliases: |
DDX30, RETCOR |
Location: |
Chr.3:47802909-47850196 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834679 | Chr.3:47707985 - 47882516 on Build GRCh38 | Loss | DHX30 MIR1226 SMARCC1 MAP4 |
nsv508916 | Chr.3:47753956 - 47883831 on Build GRCh38 | Insertion | DHX30 MIR1226 SMARCC1 MAP4 |
nsv1006560 | Chr.3:47825971 - 47995541 on Build GRCh38 | Gain | DHX30 MIR1226 MAP4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |