Assay Details
Target Gene Details
Entrez Gene ID: | 9819 |
Gene Name: | TSC22 domain family member 2 |
Gene Aliases: |
TILZ4a, TILZ4b, TILZ4c |
Location: |
Chr.3:150408268-150460119 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TSC22D2 | NM_001303264.1 | NP_001290193.1 | ||
NM_014779.3 | NP_055594.1 | |||
NR_130136.1 | ||||
XM_011513337.2 | XP_011511639.1 | |||
AB014569.1 | ||||
AF201290.1 | AAG41223.1 | |||
AF201291.1 | AAG41224.1 | |||
AF201292.1 | AAG41225.1 | |||
AK297383.1 | ||||
AK297748.1 | ||||
BC044643.1 | AAH44643.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv829755 | Chr.3:150274236 - 150438214 on Build GRCh38 | Loss | LINC01214 TSC22D2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |