Assay Details
Target Gene Details
Entrez Gene ID: | 90288 |
Gene Name: | EF-hand calcium binding domain 12 |
Gene Aliases: |
C3orf25 |
Location: |
Chr.3:129401321-129428651 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EFCAB12 | NM_207307.2 | NP_997190.1 | ||
XM_011513293.2 | XP_011511595.1 | |||
XM_011513294.2 | XP_011511596.1 | |||
XM_011513295.2 | XP_011511597.1 | |||
XM_011513297.2 | XP_011511599.1 | |||
AK127968.1 | ||||
BC066975.1 | AAH66975.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2725901 | Chr.3:129380821 - 129487939 on Build GRCh38 | Deletion | SNORA7B RPL32P3 MBD4 EFCAB12 IFT122 |
esv3597741 | Chr.3:129383049 - 129486193 on Build GRCh38 | Loss | SNORA7B RPL32P3 MBD4 EFCAB12 IFT122 |
nsv591668 | Chr.3:129403449 - 129490335 on Build GRCh38 | Gain | MBD4 EFCAB12 IFT122 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |