Assay Details
Target Gene Details
Entrez Gene ID: | 5274 |
Gene Name: | serpin family I member 1 |
Gene Aliases: |
PI12, neuroserpin |
Location: |
Chr.3:167735335-167825569 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SERPINI1 | NM_001122752.1 | NP_001116224.1 | ||
NM_005025.4 | NP_005016.1 | |||
XM_017006618.1 | XP_016862107.1 | |||
AK290082.1 | ||||
BC018043.1 | AAH18043.1 | |||
CR627434.1 | CAH10520.1 | |||
DA270615.1 | ||||
DA359846.1 | ||||
Z81326.1 | CAB03626.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |