Assay Details
Target Gene Details
Entrez Gene ID: | 8419 |
Gene Name: | beaded filament structural protein 2 |
Gene Aliases: |
CP47, CP49, CTRCT12, LIFL-L, PHAKOSIN |
Location: |
Chr.3:133397882-133475212 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BFSP2 | NM_003571.3 | NP_003562.1 | ||
XM_017007315.1 | XP_016862804.1 | |||
BC113518.1 | ||||
BC113520.1 | ||||
U48224.1 | AAC50414.1 |
Target Gene Details
Entrez Gene ID: | 85003 |
Gene Name: | BFSP2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:133426402-133491146 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BFSP2-AS1 | NR_135276.1 | |||
NR_135277.1 | ||||
NR_135278.1 | ||||
AA746278.1 | ||||
HY096628.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv4903n100 | Chr.3:133418820 - 133467964 on Build GRCh38 | Loss | BFSP2 BFSP2-AS1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |