Assay Details
Target Gene Details
Entrez Gene ID: | 6280 |
Gene Name: | S100 calcium binding protein A9 |
Gene Aliases: |
60B8AG, CAGB, CFAG, CGLB, L1AG, LIAG, MAC387, MIF, MRP14, NIF, P14 |
Location: |
Chr.1:153357854-153361027 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
S100A9 | NM_002965.3 | NP_002956.1 | ||
AF086362.1 | ||||
AK311882.1 | ||||
BC047681.2 | AAH47681.1 | |||
CR542207.1 | CAG47003.1 | |||
CR542224.1 | CAG47020.1 | |||
M26311.1 | AAA68480.1 | |||
X06233.1 | CAA29579.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1002705 | Chr.1:153335376 - 153553113 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)