Assay Details
Target Gene Details
Entrez Gene ID: | 116228 |
Gene Name: | COX20, cytochrome c oxidase assembly factor |
Gene Aliases: |
FAM36A |
Location: |
Chr.1:244835306-244845063 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
COX20 | NM_001312871.1 | NP_001299800.1 | ||
NM_001312872.1 | NP_001299801.1 | |||
NM_001312873.1 | NP_001299802.1 | |||
NM_001312874.1 | NP_001299803.1 | |||
NM_198076.5 | NP_932342.1 | |||
NR_132419.1 | ||||
NR_132420.1 | ||||
NR_132421.1 | ||||
AF086182.1 | ||||
AK000866.1 | ||||
AK125259.1 | ||||
AK130498.1 | ||||
AK300760.1 | ||||
BC018519.1 | AAH18519.1 | |||
BC062419.1 | AAH62419.1 | |||
BC095486.1 | AAH95486.1 | |||
BG708903.1 | ||||
BQ189361.1 | ||||
DA157561.1 | ||||
DA164934.1 | ||||
DA752002.1 | ||||
DB189037.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv549493 | Chr.1:244721983 - 244856983 on Build GRCh38 | Gain |
![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)