Assay Details
Target Gene Details
Entrez Gene ID: | 23266 |
Gene Name: | adhesion G protein-coupled receptor L2 |
Gene Aliases: |
CIRL2, CL2, LEC1, LPHH1, LPHN2 |
Location: |
Chr.1:81306155-81992738 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ADGRL2 | XM_017000782.1 | XP_016856271.1 | ||
AK123422.1 |
Target Gene Details
Entrez Gene ID: | 101927434 |
Gene Name: | uncharacterized LOC101927434 |
Gene Aliases: |
- |
Location: |
Chr.1:81513880-81557702 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC101927434 | NR_125943.1 | |||
BX092247.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1013276 | Chr.1:81538064 - 81604844 on Build GRCh38 | Gain |
![]() ![]() |
esv2763570 | Chr.1:81470555 - 81576665 on Build GRCh38 | Gain |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)