Assay Details
Target Gene Details
Entrez Gene ID: | 55225 |
Gene Name: | ribonucleoprotein, PTB binding 2 |
Gene Aliases: |
- |
Location: |
Chr.1:64744979-64833232 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RAVER2 | NM_018211.3 | NP_060681.2 | ||
XM_006710738.3 | XP_006710801.2 | |||
XM_011541706.2 | XP_011540008.1 | |||
AB046799.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3577903 | Chr.1:64771671 - 64776350 on Build GRCh38 | Loss | RAVER2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |