Assay Details
Target Gene Details
Entrez Gene ID: | 55509 |
Gene Name: | basic leucine zipper ATF-like transcription factor 3 |
Gene Aliases: |
JDP1, JUNDM1, SNFT |
Location: |
Chr.1:212686417-212699985 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BATF3 | NM_018664.2 | NP_061134.1 | ||
AF255346.1 | AAF73966.1 | |||
AK093671.1 | ||||
BC117489.1 | ||||
BC117491.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv549139 | Chr.1:212158952 - 212703560 on Build GRCh38 | Gain | LOC101929565 SNORA16B FAM71A NENF LOC101929541 BATF3 ATF3 PPP2R5A TMEM206 |
nsv4465 | Chr.1:212665368 - 212699073 on Build GRCh38 | Insertion | BATF3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |