Assay Details
Target Gene Details
Entrez Gene ID: | 10327 |
Gene Name: | aldo-keto reductase family 1 member A1 |
Gene Aliases: |
ALDR1, ALR, ARM, DD3, HEL-S-6 |
Location: |
Chr.1:45550779-45570051 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AKR1A1 | NM_001202413.1 | NP_001189342.1 | ||
NM_001202414.1 | NP_001189343.1 | |||
NM_006066.3 | NP_006057.1 | |||
NM_153326.2 | NP_697021.1 | |||
XM_011540491.2 | XP_011538793.1 | |||
XM_011540492.2 | XP_011538794.1 | |||
AK293083.1 | ||||
AK307068.1 | ||||
AK307237.1 | ||||
AK307288.1 | ||||
AK311175.1 | ||||
AU142853.1 | ||||
BC000670.2 | AAH00670.1 | |||
BC005394.1 | AAH05394.1 | |||
BI549149.1 | ||||
BT007003.1 | AAP35649.1 | |||
CR457010.1 | CAG33291.1 | |||
CX788871.1 | ||||
DA550374.1 | ||||
EU668319.1 | ||||
EU794588.1 | ||||
J04794.1 | AAA51711.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3893512 | Chr.1:45476180 - 46068871 on Build GRCh38 | Gain | PRDX1 TESK2 IPP NASP MAST2 GPBP1L1 RPS15AP10 AKR1A1 TMEM69 MMACHC PIK3R3 CCDC163P CCDC17 |
nsv829726 | Chr.1:45431909 - 45637514 on Build GRCh38 | Loss | AKR1A1 PRDX1 TESK2 NASP MMACHC GPBP1L1 CCDC163P CCDC17 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |