Assay Details
Target Gene Details
Entrez Gene ID: | 864 |
Gene Name: | runt related transcription factor 3 |
Gene Aliases: |
AML2, CBFA3, PEBP2aC |
Location: |
Chr.1:24899511-24965158 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RUNX3 | NM_001031680.2 | NP_001026850.1 | ||
NM_001320672.1 | NP_001307601.1 | |||
NM_004350.2 | NP_004341.1 | |||
XM_005246024.4 | XP_005246081.1 | |||
XM_011542351.1 | XP_011540653.1 | |||
AK300056.1 | ||||
AK301044.1 | ||||
BC013362.2 | AAH13362.1 | |||
X79550.2 | CAA56093.2 | |||
Z35278.1 | CAA84541.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv508581 | Chr.1:24884488 - 24968228 on Build GRCh38 | Deletion |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)