Assay Details
Target Gene Details
Entrez Gene ID: | 271 |
Gene Name: | adenosine monophosphate deaminase 2 |
Gene Aliases: |
PCH9, SPG63 |
Location: |
Chr.1:109619813-109632055 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AMPD2 | NM_001257360.1 | NP_001244289.1 | ||
NM_001308170.1 | NP_001295099.1 | |||
NM_004037.7 | NP_004028.3 | |||
NM_139156.3 | NP_631895.1 | |||
AK025706.1 | ||||
AK302939.1 | ||||
AK308907.1 | ||||
AK314494.1 | ||||
BC007711.2 | AAH07711.1 | |||
BC075844.1 | AAH75844.1 | |||
DA558264.1 | ||||
U16267.1 | AAC50306.1 | |||
U16268.1 | AAC50307.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv2299 | Chr.1:109610296 - 109662341 on Build GRCh38 | Deletion | AMPD2 GNAT2 GSTM4 |
dgv253n100 | Chr.1:109613590 - 109700319 on Build GRCh38 | Gain | GSTM1 AMPD2 GNAT2 GSTM4 GSTM2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |