Assay Details
Target Gene Details
Entrez Gene ID: | 79679 |
Gene Name: | V-set domain containing T cell activation inhibitor 1 |
Gene Aliases: |
B7-H4, B7H4, B7S1, B7X, B7h.5, PRO1291, VCTN1 |
Location: |
Chr.1:117143587-117210992 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
VTCN1 | NM_001253849.1 | NP_001240778.1 | ||
NM_001253850.1 | NP_001240779.1 | |||
NM_024626.3 | NP_078902.2 | |||
NR_045603.1 | ||||
NR_045604.1 | ||||
XM_011542143.2 | XP_011540445.2 | |||
XM_017002335.1 | XP_016857824.1 | |||
AK026071.1 | BAB15349.1 | |||
AK225413.1 | ||||
AK303466.1 | ||||
AK310812.1 | ||||
AY280972.1 | AAP37283.1 | |||
AY346100.1 | AAQ24206.1 | |||
AY358352.1 | AAQ88718.1 | |||
AY442303.1 | AAS13400.1 | |||
BC065717.1 | AAH65717.1 | |||
BC074729.2 | AAH74729.1 | |||
BX648021.1 | ||||
DB198367.1 | ||||
DC347131.1 | ||||
DQ103757.1 | AAZ17406.1 | |||
DQ836392.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1007386 | Chr.1:117162537 - 117179558 on Build GRCh38 | Loss | VTCN1 |
nsv1009706 | Chr.1:117049372 - 117442154 on Build GRCh38 | Gain | TRIM45 VTCN1 LOC101929099 LINC01525 TTF2 MAN1A2 MIR942 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |