Assay Details
Target Gene Details
Entrez Gene ID: | 58985 |
Gene Name: | interleukin 22 receptor subunit alpha 1 |
Gene Aliases: |
CRF2-9, IL22R, IL22R1 |
Location: |
Chr.1:24119771-24143285 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
IL22RA1 | NM_021258.3 | NP_067081.2 | ||
XM_011541882.1 | XP_011540184.1 | |||
AF286095.1 | AAG22073.1 | |||
AK226001.1 | ||||
AK292204.1 | ||||
AK304454.1 | ||||
AK308522.1 | ||||
AK313971.1 | ||||
BC029273.1 | AAH29273.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |