Assay Details
Target Gene Details
Entrez Gene ID: | 23254 |
Gene Name: | kazrin, periplakin interacting protein |
Gene Aliases: |
KAZ |
Location: |
Chr.1:13893387-15118048 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KAZN | XM_005245795.4 | XP_005245852.1 | ||
XM_011541074.2 | XP_011539376.1 | |||
XM_011541080.2 | XP_011539382.1 | |||
XM_017000768.1 | XP_016856257.1 | |||
XM_017000769.1 | XP_016856258.1 | |||
XM_017000770.1 | XP_016856259.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3577742 | Chr.1:13881586 - 13901995 on Build GRCh38 | Loss | KAZN |
nsv545567 | Chr.1:13558784 - 14026576 on Build GRCh38 | Gain | PDPN KAZN PRDM2 |
esv3585267 | Chr.1:13896407 - 13901691 on Build GRCh38 | Loss | KAZN |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |