Assay Details
Target Gene Details
Entrez Gene ID: | 375061 |
Gene Name: | family with sequence similarity 89 member A |
Gene Aliases: |
C1orf153 |
Location: |
Chr.1:231018958-231040249 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FAM89A | NM_198552.2 | NP_940954.1 | ||
BC009447.1 | AAH09447.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3589093 | Chr.1:230482037 - 231277190 on Build GRCh38 | Gain | C1orf131 TRIM67 MIR1182 CAPN9 COG2 TTC13 AGT FAM89A LOC101927604 LOC149373 LOC101927553 GNPAT C1orf198 ARV1 |
nsv1011066 | Chr.1:230940432 - 231242850 on Build GRCh38 | Gain | C1orf131 TRIM67 MIR1182 GNPAT TTC13 ARV1 FAM89A LOC149373 |
nsv1000488 | Chr.1:230799658 - 231269735 on Build GRCh38 | Gain | C1orf131 TRIM67 MIR1182 CAPN9 GNPAT C1orf198 TTC13 ARV1 FAM89A LOC101927604 LOC149373 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |