Assay Details
Target Gene Details
Entrez Gene ID: | 50717 |
Gene Name: | DDB1 and CUL4 associated factor 8 |
Gene Aliases: |
GAN2, H326, WDR42A |
Location: |
Chr.1:160215715-160262560 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DCAF8 | NM_015726.3 | NP_056541.2 | ||
NR_028103.1 | ||||
NR_028104.1 | ||||
AK026481.1 | ||||
AK223475.1 | BAD97195.1 | |||
AK303188.1 | ||||
AK312690.1 | ||||
BC098271.1 | AAH98271.1 | |||
BC099709.1 | AAH99709.1 | |||
BC099846.1 | AAH99846.1 | |||
CR749801.1 | CAH18661.1 | |||
U06631.1 | AAA16607.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Additional Information:
For this assay, SNP(s) [rs74125522] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)