Assay Details
Target Gene Details
Entrez Gene ID: | 6301 |
Gene Name: | seryl-tRNA synthetase |
Gene Aliases: |
SERRS, SERS |
Location: |
Chr.1:109213893-109238182 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SARS | NM_006513.3 | NP_006504.2 | ||
XM_006710813.2 | XP_006710876.1 | |||
AK222677.1 | BAD96397.1 | |||
AK312771.1 | ||||
BC000716.1 | AAH00716.1 | |||
BC009390.2 | AAH09390.1 | |||
BC111398.1 | ||||
BP363347.1 | ||||
D49914.1 | BAA95602.1 | |||
X91257.1 | CAA62635.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1008458 | Chr.1:109097053 - 109231404 on Build GRCh38 | Gain | KIAA1324 SARS SCARNA2 C1orf194 |
nsv1012267 | Chr.1:109168205 - 109279887 on Build GRCh38 | Gain | CELSR2 PSRC1 KIAA1324 SARS |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |