Assay Details
Target Gene Details
Entrez Gene ID: | 2701 |
Gene Name: | gap junction protein alpha 4 |
Gene Aliases: |
CX37 |
Location: |
Chr.1:34792958-34795747 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GJA4 | NM_002060.2 | NP_002051.2 | ||
XM_005270750.2 | XP_005270807.1 | |||
XM_017001043.1 | XP_016856532.1 | |||
AK291563.1 | ||||
AK313477.1 | ||||
BC027889.1 | AAH27889.1 | |||
BC072389.1 | AAH72389.1 | |||
M96789.2 | AAA52558.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1012995 | Chr.1:34786068 - 34884653 on Build GRCh38 | Gain | SMIM12 DLGAP3 GJA4 GJB3 |
esv1003104 | Chr.1:34790757 - 34795666 on Build GRCh38 | Deletion | GJA4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |