Assay Details
Target Gene Details
Entrez Gene ID: | 2328 |
Gene Name: | flavin containing monooxygenase 3 |
Gene Aliases: |
FMOII, TMAU, dJ127D3.1 |
Location: |
Chr.1:171090873-171117819 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FMO3 | NM_001002294.2 | NP_001002294.1 | ||
NM_001319173.1 | NP_001306102.1 | |||
NM_001319174.1 | NP_001306103.1 | |||
NM_006894.5 | NP_008825.4 | |||
XM_011509345.2 | XP_011507647.1 | |||
AK223166.1 | BAD96886.1 | |||
AK296105.1 | ||||
AK298406.1 | ||||
AK313197.1 | ||||
BC032016.1 | AAH32016.1 | |||
BM453951.1 | ||||
KU178077.1 | ||||
KU178078.1 | ||||
M83772.1 | AAA86284.1 | |||
Z47552.1 | CAA87632.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv28731 | Chr.1:171046945 - 171757821 on Build GRCh38 | Gain+Loss | FMO6P PRRC2C FMO4 TOP1P1 VAMP4 MIR1295A FMO1 MIR1295B MYOC FMO2 FMO3 MROH9 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |