Assay Details
Target Gene Details
Entrez Gene ID: | 26052 |
Gene Name: | dynamin 3 |
Gene Aliases: |
Dyna III |
Location: |
Chr.1:171841478-172418466 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831893 | Chr.1:171833310 - 172015860 on Build GRCh38 | Loss | DNM3 DNM3-IT1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |