Assay Details
Target Gene Details
Entrez Gene ID: | 100289178 |
Gene Name: | GNG12 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.1:67832288-68202987 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GNG12-AS1 | NR_040077.1 | |||
AK124028.1 | BAC85755.1 |
Target Gene Details
Entrez Gene ID: | 79971 |
Gene Name: | wntless Wnt ligand secretion mediator |
Gene Aliases: |
C1orf139, EVI, GPR177, MRP, mig-14 |
Location: |
Chr.1:68098459-68233000 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
WLS | NM_001002292.3 | NP_001002292.3 | ||
NM_001193334.1 | NP_001180263.1 | |||
NM_024911.6 | NP_079187.3 | |||
XM_011542191.2 | XP_011540493.1 | |||
XM_011542192.2 | XP_011540494.1 | |||
XM_017002390.1 | XP_016857879.1 | |||
AI217373.1 | ||||
AK026744.1 | BAB15540.1 | |||
AK074583.1 | BAC11072.1 | |||
AK074984.1 | BAC11333.1 | |||
AK301613.1 | ||||
AK309779.1 | ||||
AY359035.1 | AAQ89394.1 | |||
BC007211.1 | AAH07211.2 | |||
BC110826.1 | AAI10827.1 | |||
BC137109.1 | ||||
BC137113.1 | ||||
BX538320.1 | CAD98094.1 | |||
BX648748.1 | ||||
CA438784.1 | ||||
DQ323735.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv546448 | Chr.1:68072585 - 68177408 on Build GRCh38 | Gain |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)