Assay Details
Target Gene Details
Entrez Gene ID: | 3766 |
Gene Name: | potassium voltage-gated channel subfamily J member 10 |
Gene Aliases: |
BIRK-10, KCNJ13-PEN, KIR1.2, KIR4.1, SESAME |
Location: |
Chr.1:160037467-160070261 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KCNJ10 | NM_002241.4 | NP_002232.2 | ||
AK312383.1 | ||||
BC034036.1 | AAH34036.2 | |||
BC131627.1 | ||||
DA958759.1 | ||||
U52155.1 | AAB07046.1 |
Target Gene Details
Entrez Gene ID: | 3765 |
Gene Name: | potassium voltage-gated channel subfamily J member 9 |
Gene Aliases: |
GIRK3, KIR3.3 |
Location: |
Chr.1:160055001-160090563 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KCNJ9 | XM_017001247.1 | XP_016856736.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv3199 | Chr.1:160033106 - 160064505 on Build GRCh38 | Insertion | KCNJ10 KCNJ9 |
nsv520189 | Chr.1:160041722 - 160125835 on Build GRCh38 | Loss | ATP1A2 KCNJ10 IGSF8 KCNJ9 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |