Assay Details
Target Gene Details
Entrez Gene ID: | 1806 |
Gene Name: | dihydropyrimidine dehydrogenase |
Gene Aliases: |
DHP, DHPDHASE, DPD |
Location: |
Chr.1:97077743-97921059 on Build GRCh38 |
Assay Gene Location: | Within Intron 23 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DPYD | NM_000110.3 | NP_000101.2 | ||
XM_005270562.3 | XP_005270619.2 | |||
XM_017000507.1 | XP_016855996.1 | |||
XM_017000508.1 | XP_016855997.1 | |||
XM_017000509.1 | XP_016855998.1 | |||
XM_017000510.1 | XP_016855999.1 | |||
AB003063.1 | BAA89789.1 | |||
AK291217.1 | ||||
BC131777.1 | ||||
BC131778.1 | ||||
U09178.1 | AAA57474.1 | |||
U20938.1 | AAB51366.1 |
Target Gene Details
Entrez Gene ID: | 100873932 |
Gene Name: | DPYD antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.1:97095923-97322955 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DPYD-AS1 | NR_046590.1 | |||
CF529923.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv830692 | Chr.1:97035936 - 97159597 on Build GRCh38 | Gain+Loss |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)