Assay Details
Target Gene Details
Entrez Gene ID: | 60313 |
Gene Name: | GC-rich promoter binding protein 1 like 1 |
Gene Aliases: |
SP192 |
Location: |
Chr.1:45627304-45686691 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GPBP1L1 | NM_021639.4 | NP_067652.1 | ||
XM_017002019.1 | XP_016857508.1 | |||
AF177334.1 | AAG17978.1 | |||
AK024972.1 | BAB15047.1 | |||
AK225197.1 | ||||
AL834194.1 | CAD38884.1 | |||
BC015713.1 | AAH15713.1 | |||
BC022218.1 | AAH22218.1 | |||
BC052257.1 | AAH52257.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1014128 | Chr.1:45631998 - 45928868 on Build GRCh38 | Gain | IPP GPBP1L1 TMEM69 RPS15AP10 MAST2 |
esv3893512 | Chr.1:45476180 - 46068871 on Build GRCh38 | Gain | CCDC17 MMACHC NASP CCDC163P RPS15AP10 MAST2 TESK2 IPP GPBP1L1 PIK3R3 PRDX1 AKR1A1 TMEM69 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |