Assay Details
Target Gene Details
Entrez Gene ID: | 8672 |
Gene Name: | eukaryotic translation initiation factor 4 gamma 3 |
Gene Aliases: |
eIF-4G 3, eIF4G 3, eIF4GII |
Location: |
Chr.1:20806292-21177320 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1012238 | Chr.1:20972870 - 21040264 on Build GRCh38 | Loss |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)