Assay Details
Target Gene Details
Entrez Gene ID: | 374977 |
Gene Name: | maestro heat like repeat family member 7 |
Gene Aliases: |
C1orf175, HEATR8 |
Location: |
Chr.1:54641740-54710267 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MROH7 | NM_001039464.3 | NP_001034553.2 | ||
NM_001291332.1 | NP_001278261.1 | |||
NR_026782.2 | ||||
NR_111931.1 | ||||
AK093272.1 | BAC04116.1 | |||
AK301894.1 | ||||
AK302446.1 | ||||
AK302703.1 | ||||
AL704849.1 | ||||
DB040876.1 | ||||
DB457600.2 |
Target Gene Details
Entrez Gene ID: | 100527960 |
Gene Name: | MROH7-TTC4 readthrough (NMD candidate) |
Gene Aliases: |
HEATR8-TTC4 |
Location: |
Chr.1:54641740-54742657 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MROH7-TTC4 | NR_037639.2 | |||
NR_037640.2 | ||||
NR_037641.2 | ||||
BX648088.1 | ||||
CR749830.1 | CAH18687.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1004778 | Chr.1:54635180 - 54764639 on Build GRCh38 | Gain | MROH7-TTC4 MROH7 PARS2 TTC4 |
nsv1000673 | Chr.1:54630765 - 54818045 on Build GRCh38 | Gain | MROH7-TTC4 LEXM TTC22 MROH7 PARS2 ACOT11 TTC4 |
nsv546266 | Chr.1:54404143 - 54887697 on Build GRCh38 | Gain | MROH7-TTC4 LEXM DHCR24 TTC22 MROH7 FAM151A SSBP3 PARS2 ACOT11 TTC4 |
dgv39e212 | Chr.1:54647335 - 54655840 on Build GRCh38 | Loss | MROH7-TTC4 MROH7 |
nsv461594 | Chr.1:54593622 - 54757517 on Build GRCh38 | Gain | MROH7-TTC4 MROH7 FAM151A PARS2 ACOT11 TTC4 |
dgv38e212 | Chr.1:54627539 - 54817638 on Build GRCh38 | Gain | MROH7-TTC4 LEXM TTC22 MROH7 PARS2 ACOT11 TTC4 |
nsv817614 | Chr.1:54580862 - 55735351 on Build GRCh38 | Gain | MROH7-TTC4 TMEM61 DHCR24 LOC100507634 TTC22 FAM151A MIR4422 BSND LEXM PCSK9 MROH7 PARS2 ACOT11 TTC4 USP24 |
nsv910 | Chr.1:54605786 - 54663322 on Build GRCh38 | Deletion | MROH7-TTC4 MROH7 FAM151A ACOT11 |
nsv829904 | Chr.1:54548613 - 54721384 on Build GRCh38 | Loss | MROH7-TTC4 MROH7 FAM151A ACOT11 TTC4 |