Assay Details
Target Gene Details
Entrez Gene ID: | 3140 |
Gene Name: | major histocompatibility complex, class I-related |
Gene Aliases: |
HLALS |
Location: |
Chr.1:181033379-181061938 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MR1 | NM_001194999.1 | NP_001181928.1 | ||
NM_001195000.1 | NP_001181929.1 | |||
NM_001195035.1 | NP_001181964.1 | |||
NM_001310213.1 | NP_001297142.1 | |||
NM_001531.2 | NP_001522.1 | |||
XM_006711286.3 | XP_006711349.1 | |||
XM_011509463.2 | XP_011507765.1 | |||
AF010446.1 | AAD01442.1 | |||
AF010447.1 | AAD01443.1 | |||
AF031469.1 | AAD01933.1 | |||
AJ249778.1 | CAB77667.1 | |||
AK222910.1 | BAD96630.1 | |||
AK290374.1 | ||||
AK304645.1 | ||||
BC012485.1 | AAH12485.1 | |||
BX415555.2 | ||||
DC297418.1 | ||||
U22963.1 | AAC50174.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2719940 | Chr.1:180969855 - 181062846 on Build GRCh38 | Deletion | MR1 STX6 |
esv3890804 | Chr.1:180703656 - 181043092 on Build GRCh38 | Gain | KIAA1614 MR1 XPR1 STX6 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |