Assay Details
Target Gene Details
Entrez Gene ID: | 60676 |
Gene Name: | pappalysin 2 |
Gene Aliases: |
PAPP-A2, PAPP-E, PAPPE, PLAC3 |
Location: |
Chr.1:176463159-176845601 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PAPPA2 | NM_020318.2 | NP_064714.2 | ||
NM_021936.2 | NP_068755.2 | |||
XM_011509857.1 | XP_011508159.1 | |||
XM_017002023.1 | XP_016857512.1 | |||
XM_017002024.1 | XP_016857513.1 | |||
AF342989.1 | AAL17779.1 | |||
AF342990.1 | AAL17780.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831970 | Chr.1:176417026 - 176600071 on Build GRCh38 | Gain |
![]() |
nsv1004658 | Chr.1:176443038 - 176491284 on Build GRCh38 | Gain |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)