Assay Details
Target Gene Details
Entrez Gene ID: | 10451 |
Gene Name: | vav guanine nucleotide exchange factor 3 |
Gene Aliases: |
- |
Location: |
Chr.1:107571160-107964923 on Build GRCh38 |
Assay Gene Location: | Within Intron 30 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
VAV3 | NM_001079874.1 | NP_001073343.1 | ||
NM_006113.4 | NP_006104.4 | |||
XM_005270360.2 | XP_005270417.1 | |||
XM_017000053.1 | XP_016855542.1 | |||
XM_017000055.1 | XP_016855544.1 | |||
XM_017000056.1 | XP_016855545.1 | |||
AF067817.1 | AAC79695.1 | |||
AF118886.1 | AAD20348.1 | |||
AF118887.1 | AAD20349.1 | |||
AK296537.1 | ||||
AK300474.1 | ||||
AK304088.1 | ||||
AK316295.1 | ||||
BC143969.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv830993 | Chr.1:107441433 - 107613979 on Build GRCh38 | Gain | NTNG1 VAV3 |
dgv248n100 | Chr.1:107385172 - 108286989 on Build GRCh38 | Gain | NBPF4 NTNG1 MIR7852 VAV3 SLC25A24 VAV3-AS1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |