Assay Details
Target Gene Details
Entrez Gene ID: | 8412 |
Gene Name: | breast cancer anti-estrogen resistance 3 |
Gene Aliases: |
NSP2, SH2D3B |
Location: |
Chr.1:93561741-93847150 on Build GRCh38 |
Assay Gene Location: | Within Intron 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BCAR3 | NM_001261408.1 | NP_001248337.1 | ||
NM_001261409.1 | NP_001248338.1 | |||
NM_001261410.1 | NP_001248339.1 | |||
NM_001308251.1 | NP_001295180.1 | |||
NM_003567.3 | NP_003558.1 | |||
XM_011542251.2 | XP_011540553.1 | |||
XM_017002480.1 | XP_016857969.1 | |||
AF124250.1 | AAD28245.1 | |||
AK027785.1 | ||||
AK055442.1 | ||||
AK301570.1 | ||||
AL833121.1 | ||||
AY358996.1 | AAQ89355.1 | |||
BC039895.1 | AAH39895.1 | |||
BQ015705.1 | ||||
U92715.1 | AAC39777.1 |
Target Gene Details
Entrez Gene ID: | 54874 |
Gene Name: | formin binding protein 1 like |
Gene Aliases: |
C1orf39, TOCA1 |
Location: |
Chr.1:93448067-93569671 on Build GRCh38 |
Assay Gene Location: | Within Intron 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FNBP1L | XM_011541625.2 | XP_011539927.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1011853 | Chr.1:93500893 - 93573448 on Build GRCh38 | Loss | FNBP1L BCAR3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |