Assay Details
Target Gene Details
Entrez Gene ID: | 26135 |
Gene Name: | SERPINE1 mRNA binding protein 1 |
Gene Aliases: |
CGI-55, CHD3IP, HABP4L, PAI-RBP1, PAIRBP1 |
Location: |
Chr.1:67407810-67430440 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SERBP1 | NM_001018067.1 | NP_001018077.1 | ||
NM_001018068.1 | NP_001018078.1 | |||
NM_001018069.1 | NP_001018079.1 | |||
NM_015640.3 | NP_056455.3 | |||
AF151813.1 | AAD34050.1 | |||
AK074970.1 | BAC11324.1 | |||
AK130606.1 | ||||
AK222955.1 | BAD96675.1 | |||
AK223138.1 | BAD96858.1 | |||
AK312256.1 | ||||
AL080119.1 | CAB45718.1 | |||
AY032853.1 | AAK51130.1 | |||
BC002488.2 | AAH02488.1 | |||
BC003049.1 | AAH03049.1 | |||
BC008045.2 | AAH08045.1 | |||
BC017449.1 | AAH17449.1 | |||
BC019273.1 | AAH19273.1 | |||
BC020555.1 | AAH20555.1 | |||
BC026916.1 | AAH26916.1 | |||
BX647139.1 | CAH56188.1 | |||
CR457383.1 | CAG33664.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv546447 | Chr.1:67356694 - 67442961 on Build GRCh38 | Loss |
![]() ![]() |
nsv1254 | Chr.1:67394481 - 67428346 on Build GRCh38 | Insertion |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)