Assay Details
Target Gene Details
Entrez Gene ID: | 28982 |
Gene Name: | feline leukemia virus subgroup C cellular receptor 1 |
Gene Aliases: |
AXPC1, FLVCR, MFSD7B, PCA, PCARP |
Location: |
Chr.1:212858255-212899363 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FLVCR1 | NM_014053.3 | NP_054772.1 | ||
XM_011509446.2 | XP_011507748.1 | |||
AF118637.1 | AAD45243.1 | |||
AK001419.1 | BAA91679.1 | |||
AK314481.1 | ||||
BC048312.1 | AAH48312.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1007129 | Chr.1:212863902 - 212909919 on Build GRCh38 | Gain | FLVCR1 |
dgv572n100 | Chr.1:212856472 - 212899671 on Build GRCh38 | Gain | FLVCR1 FLVCR1-AS1 |
dgv571n100 | Chr.1:212831823 - 212896221 on Build GRCh38 | Gain | FLVCR1 SPATA45 FLVCR1-AS1 |
nsv1008383 | Chr.1:212852204 - 212941552 on Build GRCh38 | Gain | VASH2 FLVCR1 FLVCR1-AS1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |