Assay Details
Target Gene Details
Entrez Gene ID: | 148327 |
Gene Name: | cAMP responsive element binding protein 3 like 4 |
Gene Aliases: |
AIBZIP, ATCE1, CREB3, CREB4, JAL, hJAL |
Location: |
Chr.1:153967679-153974364 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CREB3L4 | NM_001255978.1 | NP_001242907.1 | ||
NM_001255979.1 | NP_001242908.1 | |||
NM_001255980.1 | NP_001242909.1 | |||
NM_001255981.1 | NP_001242910.1 | |||
NM_130898.3 | NP_570968.1 | |||
NR_045658.1 | ||||
XM_006711172.2 | XP_006711235.1 | |||
XM_017000372.1 | XP_016855861.1 | |||
AB052778.1 | BAC45035.1 | |||
AF394167.1 | AAL76113.1 | |||
AF468007.1 | AAO33070.1 | |||
AK297703.1 | ||||
AK304665.1 | ||||
AY049977.1 | AAL13157.1 | |||
BC038962.1 | AAH38962.1 | |||
BF681098.1 | ||||
BX647716.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)