Assay Details
Target Gene Details
Entrez Gene ID: | 23139 |
Gene Name: | microtubule associated serine/threonine kinase 2 |
Gene Aliases: |
MAST205, MTSSK |
Location: |
Chr.1:45786992-46036124 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MAST2 | XM_011541065.1 | XP_011539367.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3585914 | Chr.1:45718616 - 45877574 on Build GRCh38 | Gain | IPP MAST2 |
nsv1014128 | Chr.1:45631998 - 45928868 on Build GRCh38 | Gain | RPS15AP10 TMEM69 IPP GPBP1L1 MAST2 |
nsv461417 | Chr.1:45757414 - 45950676 on Build GRCh38 | Gain | MAST2 |
esv3893512 | Chr.1:45476180 - 46068871 on Build GRCh38 | Gain | TESK2 RPS15AP10 TMEM69 IPP GPBP1L1 CCDC17 CCDC163P MMACHC NASP MAST2 PIK3R3 PRDX1 AKR1A1 |
nsv524147 | Chr.1:45770426 - 45972404 on Build GRCh38 | Gain | MAST2 |
dgv159n100 | Chr.1:45706164 - 45997340 on Build GRCh38 | Gain | IPP MAST2 |
nsv1013628 | Chr.1:45755945 - 45890893 on Build GRCh38 | Gain | MAST2 |
nsv546158 | Chr.1:45750903 - 46029762 on Build GRCh38 | Gain | MAST2 |
esv3585915 | Chr.1:45720361 - 46036324 on Build GRCh38 | Gain | IPP MAST2 |
esv3577360 | Chr.1:45654763 - 45881711 on Build GRCh38 | Gain | TMEM69 IPP GPBP1L1 MAST2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |