Assay Details
Target Gene Details
Entrez Gene ID: | 28316 |
Gene Name: | cadherin 20 |
Gene Aliases: |
CDH7L3, Cdh7 |
Location: |
Chr.18:61333582-61555773 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CDH20 | NM_031891.3 | NP_114097.2 | ||
AK290190.1 | ||||
AK313809.1 | ||||
DA333316.1 | ||||
DA339323.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)