Assay Details
Target Gene Details
Entrez Gene ID: | 9480 |
Gene Name: | one cut homeobox 2 |
Gene Aliases: |
OC-2, OC2 |
Location: |
Chr.18:57435420-57491298 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ONECUT2 | NM_004852.2 | NP_004843.2 | ||
Y18198.1 | CAB38253.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)