Assay Details
Target Gene Details
Entrez Gene ID: | 101927323 |
Gene Name: | PPP4R1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.18:9614807-9619363 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PPP4R1-AS1 | NR_110773.1 | |||
NR_110774.1 | ||||
BX108181.1 | ||||
HY048010.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv458024 | Chr.18:9558242 - 9828451 on Build GRCh38 | Gain | PPP4R1-AS1 PPP4R1 RAB31 |