Assay Details
Target Gene Details
Entrez Gene ID: | 147495 |
Gene Name: | APC down-regulated 1 |
Gene Aliases: |
B7323, DRAPC1, FP7019, HHS, HTS, HYPT1 |
Location: |
Chr.18:10454628-10488701 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
APCDD1 | NM_153000.4 | NP_694545.1 | ||
XM_011525603.1 | XP_011523905.1 | |||
AB056722.1 | BAC15563.1 | |||
AB104887.1 | BAC65165.1 | |||
AF461902.1 | AAQ04816.1 | |||
AK300743.1 | ||||
AK303076.1 | ||||
BC053324.1 | AAH53324.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1059110 | Chr.18:10142986 - 10596015 on Build GRCh38 | Gain | LINC01254 NAPG APCDD1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |