Assay Details
Target Gene Details
Entrez Gene ID: | 171586 |
Gene Name: | abhydrolase domain containing 3 |
Gene Aliases: |
LABH3 |
Location: |
Chr.18:21650897-21704805 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ABHD3 | NM_001308256.1 | NP_001295185.1 | ||
NM_138340.4 | NP_612213.2 | |||
XM_011525847.2 | XP_011524149.1 | |||
XM_017025571.1 | XP_016881060.1 | |||
XM_017025572.1 | XP_016881061.1 | |||
XM_017025573.1 | XP_016881062.1 | |||
XM_017025574.1 | XP_016881063.1 | |||
AF007152.1 | AAC19155.1 | |||
AK024880.1 | ||||
AK298728.1 | ||||
BC021196.2 | AAH21196.1 | |||
BI918326.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2716850 | Chr.18:21667117 - 21674310 on Build GRCh38 | Deletion |
![]() |
esv3641939 | Chr.18:21667886 - 21690539 on Build GRCh38 | Loss |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)