Assay Details
Target Gene Details
Entrez Gene ID: | 9984 |
Gene Name: | THO complex 1 |
Gene Aliases: |
HPR1, P84, P84N5 |
Location: |
Chr.18:214348-268059 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
THOC1 | NM_005131.2 | NP_005122.2 | ||
XM_011525772.2 | XP_011524074.1 | |||
XM_011525773.1 | XP_011524075.1 | |||
XM_011525774.2 | XP_011524076.1 | |||
XM_017026103.1 | XP_016881592.1 | |||
AK055354.1 | ||||
AK225067.1 | ||||
AK225709.1 | ||||
AK314755.1 | ||||
AY573303.1 | AAT81409.1 | |||
BC010381.1 | AAH10381.1 | |||
L36529.1 | AAA53571.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv2174 | Chr.18:240074 - 285087 on Build GRCh38 | Deletion |
![]() |
dgv3303n100 | Chr.18:219990 - 290812 on Build GRCh38 | Gain |
![]() |
nsv576227 | Chr.18:52933 - 322183 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |
esv3641475 | Chr.18:153985 - 282924 on Build GRCh38 | Gain |
![]() ![]() |
nsv518642 | Chr.18:196829 - 331506 on Build GRCh38 | Gain |
![]() ![]() ![]() |
nsv1064150 | Chr.18:99036 - 321710 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |
dgv3301n100 | Chr.18:11543 - 326679 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |
nsv1160503 | Chr.18:200576 - 249255 on Build GRCh38 | Duplication |
![]() ![]() |
nsv482935 | Chr.18:10001 - 2900002 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
dgv3302n100 | Chr.18:136228 - 249310 on Build GRCh38 | Gain |
![]() ![]() |
esv3641473 | Chr.18:128077 - 323706 on Build GRCh38 | Gain |
![]() ![]() ![]() |
nsv576228 | Chr.18:69836 - 1506323 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)