Assay Details
Target Gene Details
Entrez Gene ID: | 652991 |
Gene Name: | SKI family transcriptional corepressor 2 |
Gene Aliases: |
CH18515, CORL2, FUSSEL18 |
Location: |
Chr.18:47206182-47249524 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SKOR2 | NM_001278063.1 | NP_001264992.1 | ||
XM_006722525.3 | XP_006722588.1 | |||
DW009659.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv9629 | Chr.18:46684730 - 47231140 on Build GRCh38 | Gain+Loss | KATNAL2 ST8SIA5 SKOR2 TCEB3B TCEB3C HDHD2 PIAS2 TCEB3CL2 LOC107983955 TCEB3CL IER3IP1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |