Assay Details
Target Gene Details
Entrez Gene ID: | 4200 |
Gene Name: | malic enzyme 2 |
Gene Aliases: |
ODS1 |
Location: |
Chr.18:50878604-50949795 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ME2 | NM_001168335.1 | NP_001161807.1 | ||
NM_002396.4 | NP_002387.1 | |||
AK313391.1 | ||||
AU119085.1 | ||||
BC000147.1 | AAH00147.1 | |||
BI465083.1 | ||||
BI559761.1 | ||||
DA644731.1 | ||||
M55905.1 | AAA36197.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3893102 | Chr.18:50871431 - 50953868 on Build GRCh38 | Loss | ME2 |
nsv527714 | Chr.18:50818407 - 50888556 on Build GRCh38 | Gain | ME2 MRO |
nsv9630 | Chr.18:50532107 - 50922466 on Build GRCh38 | Loss | ME2 MAPK4 MRO |
nsv576892 | Chr.18:50746815 - 50899947 on Build GRCh38 | Gain | ME2 MRO |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |