Assay Details
Target Gene Details
Entrez Gene ID: | 7050 |
Gene Name: | TGFB induced factor homeobox 1 |
Gene Aliases: |
HPE4, TGIF |
Location: |
Chr.18:3411927-3458412 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TGIF1 | NM_001278686.1 | NP_001265615.1 | ||
NM_174886.2 | NP_777480.1 | |||
BE906361.1 | ||||
BM849453.1 | ||||
CB996365.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv2190 | Chr.18:3408994 - 3442002 on Build GRCh38 | Insertion | TGIF1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |