Assay Details
Target Gene Details
Entrez Gene ID: | 5892 |
Gene Name: | RAD51 paralog D |
Gene Aliases: |
BROVCA4, R51H3, RAD51L3, TRAD |
Location: |
Chr.17:35099792-35119869 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RAD51D | NM_001142571.1 | NP_001136043.1 | ||
NM_002878.3 | NP_002869.3 | |||
NM_133629.2 | NP_598332.1 | |||
NR_037711.1 | ||||
NR_037712.1 | ||||
AB013341.1 | BAA25914.1 | |||
AB016223.1 | BAA31747.1 | |||
AB016224.1 | BAA31748.1 | |||
AB016225.1 | BAA31749.1 | |||
AB018360.1 | BAA33779.1 | |||
AB018361.1 | BAA33780.1 | |||
AB018362.1 | BAA33781.1 | |||
AB018363.1 | BAA33782.1 | |||
AF034956.1 | AAC39719.1 | |||
AK296241.1 | ||||
AK301249.1 | ||||
AK302450.1 | ||||
BC002723.2 | AAH02723.1 | |||
BC014422.1 | AAH14422.1 | |||
BI916871.1 | ||||
BX647297.1 | ||||
Y15572.1 | CAA75681.1 |
Target Gene Details
Entrez Gene ID: | 100529207 |
Gene Name: | RAD51L3-RFFL readthrough |
Gene Aliases: |
- |
Location: |
Chr.17:35009112-35121522 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RAD51L3-RFFL | NR_037714.1 | |||
AK097811.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3892998 | Chr.17:35106468 - 35295755 on Build GRCh38 | Loss | UNC45B RAD51L3-RFFL FNDC8 SLFN5 LOC105371932 SLC35G3 NLE1 RAD51D |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |