Assay Details
Target Gene Details
Entrez Gene ID: | 239 |
Gene Name: | arachidonate 12-lipoxygenase, 12S type |
Gene Aliases: |
12-LOX, 12S-LOX, LOG12 |
Location: |
Chr.17:6993791-7010736 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ALOX12 | NM_000697.2 | NP_000688.2 | ||
XM_011523780.2 | XP_011522082.1 | |||
BC069557.1 | AAH69557.1 | |||
M35418.1 | AAA60056.1 | |||
M58704.1 | AAA59523.1 | |||
M62982.1 | AAA51533.1 | |||
S68587.1 | AAD14020.1 | |||
S68588.1 | AAD14021.1 |
Target Gene Details
Entrez Gene ID: | 100506713 |
Gene Name: | ALOX12 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.17:6985123-7012334 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ALOX12-AS1 | NR_040089.1 | |||
BI559919.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2422288 | Chr.17:6789933 - 7646709 on Build GRCh38 | Deletion | CHRNB1 C17orf49 FXR2 RNASEK-C17orf49 SNORA48 SLC35G6 PHF23 FBXO39 SHBG SLC16A13 TMEM256-PLSCR3 TNK1 SPEM1 CLEC10A TEKT1 TMEM102 ALOX12 EIF4A1 SAT2 MIR195 SLC16A11 ASGR2 LOC100996842 TMEM256 MIR497HG GABARAP SLC2A4 DLG4 ACADVL ELP5 SOX15 MIR324 CTDNEP1 RNASEK ASGR1 SENP3-EIF4A1 POLR2A PLSCR3 NLGN2 SNORD10 TMEM95 TNFSF12-TNFSF13 LOC107983988 NEURL4 TNFSF13 FGF11 BCL6B ALOX12-AS1 ALOX12P2 MPDU1 DVL2 ATP1B2 SENP3 ZBTB4 SNORA67 TNFSF12 KCTD11 C17orf74 CLDN7 EIF5A GPS2 MIR497 YBX2 ACAP1 CD68 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |